3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only
Reagent for the generation of chemiluminescence.
Zhang, G.; Tang, Y.; Shang, J.; Wang, Z.; Yu, H.; Du, W.; Fu, Q. Flow-injection chemiluminescence method to detect a beta2 adrenergic agonist. Luminescence 2015, 30 (1), 102-109.
Sampaio, T. R.; Fonseca, A. Chemiluminescence detection in urethane-acrylate microfluidic devices. Sens. Actuators, B 2015, 213, 215-221.