Thermo Scientific Chemicals

4,4'-Dimethoxybenzhydrol, 98+%, Thermo Scientific Chemicals

Catalog number: A14604.09
10 g, Each
Thermo Scientific Chemicals

4,4'-Dimethoxybenzhydrol, 98+%, Thermo Scientific Chemicals

Catalog number: A14604.09
10 g, Each
Quantity
Catalog number: A14604.09
also known as A14604-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
728-87-0
Appearance (Color)
White
Melting Point (clear melt)
67.0-73.0?C
Identification (FTIR)
Conforms
Form
Powder
Assay (HPLC)
≥98.0%

Description

4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text