2,5-Di-tert-butylhydroquinone, 98+%, Thermo Scientific Chemicals
2,5-Di-tert-butylhydroquinone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Di-tert-butylhydroquinone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A14606.30
also known as A14606-30
Price (USD)/ Each
40.30
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
40.30
Add to cart
2,5-Di-tert-butylhydroquinone, 98+%, Thermo Scientific Chemicals
Catalog numberA14606.30
Price (USD)/ Each
40.30
-
Add to cart
Chemical Identifiers
CAS88-58-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline/granular powder
Assay (HPLC)≥98.0%
Appearance (Color)White to cream to pink to pale brown
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only