Large Vessel Endothelial Supplement (LVES)
Gibco™

Large Vessel Endothelial Supplement (LVES)

Gibco™ Large Vessel Endothelial Supplement (LVES) is a 50X, complete, ready-to-use media supplement optimized for use with Gibco™ Medium 200자세히 알아보기
Have Questions?
카탈로그 번호수량
A146080111 mL
카탈로그 번호 A1460801
제품 가격(KRW)
153,000
온라인 행사
Ends: 31-Mar-2026
170,000
할인액 17,000 (10%)
Each
수량:
11 mL
제품 가격(KRW)
153,000
온라인 행사
Ends: 31-Mar-2026
170,000
할인액 17,000 (10%)
Each
Gibco™ Large Vessel Endothelial Supplement (LVES) is a 50X, complete, ready-to-use media supplement optimized for use with Gibco™ Medium 200 and Medium 200PRF for culture of primary human umbilical vein endothelial cells (HUVEC), human aortic endothelial cells (HAEC), and human pulmonary artery endothelial cells (HPAEC). This blended supplement has been designed to deliver optimal cell growth and to enable superior cell performance for angiogenesis applications, such as cell migration and tube formation. LVES contains fetal bovine serum, hydrocortisone, human epidermal growth factor, basic fibroblast growth factor, heparin, and ascorbic acid.

• Perform tube formation, cell invasion, cell migration, and cell proliferation functional assays
• Achieve superior growth rates of endothelial cells compared to low serum growth supplement (LSGS)
• Expand primary large vessel endothelial cells without genomic alteration

Verified Performance in Key Angiogenesis Applications
Large Vessel Endothelial Supplement is designed to deliver optimal performance in tube formation, cell invasion, cell migration, and cell proliferation assays when used in conjunction with Medium 200, human umbilical vein endothelial cells, and Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix. These items and a detailed protocol are available in a convenient Angiogenesis Starter Kit or they can be purchased separately.

Maximum Primary Cell Growth with One Blended Supplement
Large Vessel Endothelial Supplement is a complete, ready-to-use blend of growth supplements optimized to deliver>4 x 104 HUVEC per cm2 when used with Medium 200 or Medium 200PRF. Cell phenotype is retained and verified positive for von Willebrand factor (vWf) and CD31, negative for a-actin, and positive for dil-Ac-LDL uptake via ICC.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포 유형Endothelial Cells (Umbilical Vein, Pulmonary Artery, Aortic, Large Vessel)
농도50 X
순도 또는 품질 등급Research Grade
수량11 mL
혈청 수준Standard Serum
배송 조건Dry Ice
소스Human
Human
테스트됨Growth Performance Assay
분류Animal Origin
형태Liquid
제품 유형LVES Supplement
멸균Sterile
첨가제 없음No Glutamine
Unit SizeEach
구성 및 보관
1 bottle. Store in freezer (-5 to -30°C).