Thermo Scientific Chemicals

Tin(II) chloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A14610.22
100 g, Each
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A14610.22
100 g, Each
Quantity
Catalog number: A14610.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-69-1
IUPAC Name
λ²-tin(2+) dihydrate dichloride
Molecular Formula
Cl2H4O2Sn
InChI Key
FWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Sn++]
Form
Crystalline powder or crystals and/or chunks
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Melting Point (clear melt)
41-48?C

Description

Tin(II) chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

Solubility
Soluble in dilute and concentrated hydrochloric acid, alcohol, ethyl acetate, glacial acetic acid, and sodium hydroxide solution. Decomposes in excess water.

Notes
Hygroscopic, and absorbs moisture or water from air. Incompatible with metals, strong oxidizing agents, strong bases and bromine trifluoride.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text