Thermo Scientific Chemicals

N-Phenylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: A14616.22
100 g, Each
Thermo Scientific Chemicals

N-Phenylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: A14616.22
100 g, Each
Quantity
Catalog number: A14616.22
also known as A14616-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
941-69-5
IUPAC Name
1-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Formula
C10H7NO2
InChI Key
HIDBROSJWZYGSZ-UHFFFAOYSA-N
SMILES
O=C1C=CC(=O)N1C1=CC=CC=C1
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
86.5-92.5?C
Assay (GC)
≥98.0%

Description

N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text