Thermo Scientific Chemicals

1,2-Diphenylethane, 98+%, Thermo Scientific Chemicals

Catalog number: A14617.22
100 g, Each
Thermo Scientific Chemicals

1,2-Diphenylethane, 98+%, Thermo Scientific Chemicals

Catalog number: A14617.22
100 g, Each
Quantity
Catalog number: A14617.22
also known as A14617-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-29-7
IUPAC Name
(2-phenylethyl)benzene
Molecular Formula
C14H14
InChI Key
QWUWMCYKGHVNAV-UHFFFAOYSA-N
SMILES
C(CC1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder or fused solid or chunks
Appearance (Color)
Colorless to white to pale yellow
Melting Point (clear melt)
48.0-55.0?C

Description

1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

Solubility
Soluble in ether, chloroform

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text