Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14618.22
100 g, Each
Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14618.22
100 g, Each
Quantity
Catalog number: A14618.22
also known as A14618-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
452-58-4
IUPAC Name
pyridine-2,3-diamine
Molecular Formula
C5H7N3
InChI Key
ZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILES
NC1=CC=CN=C1N
Appearance (Color)
Dark brown to brown to black
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
109.0-117.0?C

Description

2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Solubility
Soluble in water (100 g/L).

Notes
It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text