Thermo Scientific Chemicals

Isoxazole-5-carbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A14625.06
5 g, Each
Thermo Scientific Chemicals

Isoxazole-5-carbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A14625.06
5 g, Each
Quantity
Catalog number: A14625.06
also known as A14625-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62348-13-4
IUPAC Name
1,2-oxazole-5-carbonyl chloride
Molecular Formula
C4H2ClNO2
InChI Key
NASLINFISOTVJJ-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=NO1
Refractive Index
1.4965-1.5015 @ 20?C
Appearance (Color)
Clear colorless to very pale yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text