Thermo Scientific Chemicals

Thiosemicarbazide, 98%, Thermo Scientific Chemicals

Catalog number: A14630.22
100 g, Each
Thermo Scientific Chemicals

Thiosemicarbazide, 98%, Thermo Scientific Chemicals

Catalog number: A14630.22
100 g, Each
Quantity
Catalog number: A14630.22
also known as A14630-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-19-6
IUPAC Name
aminothiourea
Molecular Formula
CH5N3S
InChI Key
BRWIZMBXBAOCCF-UHFFFAOYSA-N
SMILES
NNC(N)=S
Appearance (Color)
White to pale cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

Solubility
Soluble in water, ethanol, alcohols and hot methanol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text