Thermo Scientific Chemicals

Sodium 1-decanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A14638.14
25 g, Each
Thermo Scientific Chemicals

Sodium 1-decanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A14638.14
25 g, Each
Quantity
Catalog number: A14638.14
also known as A14638-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13419-61-9
IUPAC Name
sodium decane-1-sulfonate
Molecular Formula
C10H21NaO3S
InChI Key
AIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCS([O-])(=O)=O
Form
Powder
Appearance (Color)
White
Assay (ex Sulphated Ash)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Loss on Drying
≤1.5% (105?C/constant wt.)

Description

Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text