Thermo Scientific Chemicals

Brilliant Crocein, Thermo Scientific Chemicals

Catalog number: A14642.18
50 g, Each
Thermo Scientific Chemicals

Brilliant Crocein, Thermo Scientific Chemicals

Catalog number: A14642.18
50 g, Each
Quantity
Catalog number: A14642.18
also known as A14642-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5413-75-2
IUPAC Name
disodium (8Z)-7-oxo-8-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-7,8-dihydronaphthalene-1,3-disulfonate
Molecular Formula
C22H14N4Na2O7S2
InChI Key
OJPSNKJNYKPDPH-HKWNQDJZSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(C(=NNC3=CC=C(C=C3)N=NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
lambda-max (UV-Vis)
505-515nm (in water)
Appearance (Color)
Red to red-brown
Loss on Drying
<10% (200?C/constant wt)
Specific Absorbance (E 1%/1cm)
450-700
Form
Powder

Description

Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text