Thermo Scientific Chemicals

Phenylhydrazine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A14645.22
100 g, Each
Thermo Scientific Chemicals

Phenylhydrazine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A14645.22
100 g, Each
Quantity
Catalog number: A14645.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-88-1
IUPAC Name
hydrogen phenylhydrazine chloride
Molecular Formula
C6H9ClN2
InChI Key
JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=CC=C1
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Appearance (Color)
White to cream
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (HPLC)
>98.5%

Description

Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It is used in steroid assays and as an N-protecting reagent. Its free base, phenylhydrazine is used to prepare indoles, which find application as intermediates in the synthesis of various dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It i
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text