Thermo Scientific Chemicals

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14653.14
25 g, Each
Thermo Scientific Chemicals

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14653.14
25 g, Each
Quantity
Catalog number: A14653.14
also known as A14653-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4755-77-5
IUPAC Name
ethyl oxalochloridate
Molecular Formula
C4H5ClO3
InChI Key
OWZFULPEVHKEKS-UHFFFAOYSA-N
SMILES
CCOC(=O)C(Cl)=O
Refractive Index
1.4150-1.4190 @ 20°C
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms

Description

Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text