Methyl 3,4-diaminobenzoate, 98%, Thermo Scientific Chemicals
Methyl 3,4-diaminobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3,4-diaminobenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14656.06
also known as A14656-06
Price (USD)/ Each
99.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.80
Add to cart
Methyl 3,4-diaminobenzoate, 98%, Thermo Scientific Chemicals
Catalog numberA14656.06
Price (USD)/ Each
99.80
-
Add to cart
Chemical Identifiers
CAS36692-49-6
IUPAC Namemethyl 3,4-diaminobenzoate
Molecular FormulaC8H10N2O2
InChI KeyIOPLHGOSNCJOOO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown to dark brown to dark red
Assay (GC)≥97.5%
Melting Point (clear melt)103.0-110.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only