Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.
Solubility
Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only
AEM Smedman.; IB Gustafsson. Pentadecanoic acid in serum as a marker for intake of milk fat: relations between intake of milk fat and metabolic risk factors. Am J Clin Nutr.199969 (1), 22-29.
V Dilsizian.; TM Bateman.; SR Bergmann.; R Des Prez. Metabolic imaging with β-methyl-p-[123I]-iodophenyl-pentadecanoic acid identifies ischemic memory after demand ischemia. Circulation.2005112 (1), 2169-2174.