Pentadecanoic acid, 99%, Thermo Scientific Chemicals
Pentadecanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentadecanoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A14664.18
also known as A14664-18
Price (USD)/ Each
176.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
176.00
Add to cart
Pentadecanoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA14664.18
Price (USD)/ Each
176.00
-
Add to cart
Chemical Identifiers
CAS1002-84-2
IUPAC Namepentadecanoic acid
Molecular FormulaC15H30O2
InChI KeyWQEPLUUGTLDZJY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
Melting Point (clear melt)51-55?C
FormCrystalline powder or crystals or powder or flakes and/or chunks
Identification (FTIR)Conforms
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanoic acid is a lipid used in proteomics research. It is also used as a biomarker for dietary milk intake. It can be labelled as a biomarker for myocardial fatty acid metabolism in ischemia/reperfusion studies.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only

General References:

  1. AEM Smedman.; IB Gustafsson. Pentadecanoic acid in serum as a marker for intake of milk fat: relations between intake of milk fat and metabolic risk factors. Am J Clin Nutr.199969 (1), 22-29.
  2. V Dilsizian.; TM Bateman.; SR Bergmann.; R Des Prez. Metabolic imaging with β-methyl-p-[123I]-iodophenyl-pentadecanoic acid identifies ischemic memory after demand ischemia. Circulation.2005112 (1), 2169-2174.