6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14666.06
also known as A14666-06
Price (USD)/ Each
49.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
49.20
Add to cart
6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
Catalog numberA14666.06
Price (USD)/ Each
49.20
-
Add to cart
Chemical Identifiers
CAS774-55-0
IUPAC Name1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Molecular FormulaC12H14O
InChI KeyVEPUKHYQNXSSKV-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C2CCCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5590-1.5640 @ 20?C
Assay (GC)≥96.0%
FormLiquid
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. AM Api.; RL Smith.; S Pipino.; T Marczylo. Evaluation of the oral subchronic toxicity of AHTN (7-acetyl-1, 1, 3, 4, 4, 6-hexamethyl-1, 2, 3,4-tetrahydronaphthalene) in the rat. Food and Chemical Toxicology.200542 (5), 791-801.
  2. X Zeng.; G Sheng.; Y Xiong.; J Fu. Determination of polycyclic musks in sewage sludge from Guangdong, China using GC-EI-MS. Chemosphere.200560 (6), 817-823.