3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water but soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Air Sensitive.
RUO – Research Use Only
DA Alonso.; M Fuensanta.; C Nájera. 3, 5-Bis (trifluoromethyl) phenyl sulfones in the direct Julia-Kocienski olefination. J. Org. Chem.200570 (16), 6404-6416.
YY Noh.; N Zhao.; M Caironi.; H Sirringhaus. Downscaling of self-aligned, all-printed polymer thin-film transistors. Nature nanotechnology.20072 (1), 784 - 789 .