Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)thiophenol, 98%, Thermo Scientific Chemicals

Catalog number: A14669.06
5 g, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)thiophenol, 98%, Thermo Scientific Chemicals

Catalog number: A14669.06
5 g, Each
Quantity
Catalog number: A14669.06
also known as A14669-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130783-02-7
IUPAC Name
[3,5-bis(trifluoromethyl)phenyl]sulfanide
Molecular Formula
C8H3F6S
InChI Key
KCAQWPZIMLLEAF-UHFFFAOYSA-M
SMILES
FC(F)(F)C1=CC(=CC([S-])=C1)C(F)(F)F
Form
Liquid
Refractive Index
1.4400-1.4440 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water but soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Air Sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text