2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
RUO – Research Use Only
General References:
- D Li.; J Guang.; WX Zhang.; Y Wang.; Z Xi. Zn (OTf) 2-catalyzed addition of amines to carbodiimides: efficient synthesis of guanidines and unpredicted formation of Zn-N amido species. Organic & Biomolecular chemistry. 20108 (1), 1816-1820.
- JM Casas.; J Forniés.; A Martín. Ligand structural factors in the establishment of intramolecular Pt··· H hydrogen-bridging interactions in pentafluorophenyl complexes of platinum. Journal of the Chemical Society, Dalton . 19972 (1), 1559-1564.