Thermo Scientific Chemicals

Phenylarsine oxide, 97%, Thermo Scientific Chemicals

Catalog number: A14674.06
5 g, Each
Thermo Scientific Chemicals

Phenylarsine oxide, 97%, Thermo Scientific Chemicals

Catalog number: A14674.06
5 g, Each
Quantity
Catalog number: A14674.06
also known as A14674-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
637-03-6
IUPAC Name
arsorosobenzene
Molecular Formula
C6H5AsO
InChI Key
BQVCCPGCDUSGOE-UHFFFAOYSA-N
SMILES
O=[As]C1=CC=CC=C1
Form
Powder
Assay from Supplier's CofA
≥96.0%

Description

Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

Solubility
Soluble in dimethyl sulfoxide, chloroform, benzene, ether, ethanol, 50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0 mg/ml) and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text