Thermo Scientific Chemicals

4-Aminophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14675.18
50 g, Each
Thermo Scientific Chemicals

4-Aminophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14675.18
50 g, Each
Quantity
Catalog number: A14675.18
also known as A14675-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1197-55-3
IUPAC Name
2-(4-aminophenyl)acetic acid
Molecular Formula
C8H9NO2
InChI Key
CSEWAUGPAQPMDC-UHFFFAOYSA-N
SMILES
NC1=CC=C(CC(O)=O)C=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to dark cream or yellow or pale brown
Assay (Silylated GC)
≥97.5%

Description

4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text