4-Fluorobenzyl chloride, 99%, Thermo Scientific Chemicals
4-Fluorobenzyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorobenzyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A14686.22
also known as A14686-22
Price (USD)/ Each
97.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
97.50
Add to cart
4-Fluorobenzyl chloride, 99%, Thermo Scientific Chemicals
Catalog numberA14686.22
Price (USD)/ Each
97.50
-
Add to cart
Chemical Identifiers
CAS352-11-4
IUPAC Name1-(chloromethyl)-4-fluorobenzene
Molecular FormulaC7H6ClF
InChI KeyIZXWCDITFDNEBY-UHFFFAOYSA-N
SMILESFC1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5115-1.5145 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only