Thermo Scientific™

LanthaScreen™ LRRK2 G2019S [pSer935] Cellular Assay Kit

Catalog number: A14686
Have Questions?
Thermo Scientific™

LanthaScreen™ LRRK2 G2019S [pSer935] Cellular Assay Kit

Catalog number: A14686
Have Questions?
Catalog Number
A14686
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A146861 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The LanthaScreen™ LRRK2 G2019S [pSer935] Cellular Assay Kit combines the flexibility of the BacMam gene delivery system with the robustness and power of our LanthaScreen™ TR-FRET technology. It allows researchers to interrogate phosphorylation at serine 935 on a disease-relevant mutant (G2019S) Leucine-Rich Repeat Kinase-2 (LRRK2) protein in a variety of cell backgrounds to identify LRRK2 kinase inhibitors.

This assay format provides a convenient tool for the expression of G2019S LRRK2-GFP fusion protein in the cell background of your choice. The resulting cellular assay can be used to identify LRRK2 G2019S inhibitors, aiding in Parkinson's disease and other research focuses.

This kit provides enough material to evaluate the technology in a 384-well plate; larger sizes of all reagents are available separately.

The kit includes:

• LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody, 5 μg
• BacMam LRRK2-GFP G2019S Reagent, 5 × 1 mL
• LanthaScreen™ 6x Lysis Buffer, 6 mL
• Instrument Control Terbium TR-FRET kit

A kit for interrogating wild-type LRRK2 protein is also available.

Benefits of this assay include:

More Physiologically Relevant: cell-based LRRK2 kinase activity and choice of cell background
More Convenient: compatible with High Throughput Screening (HTS); LanthaScreen™ technology reduces interference and noise
Disease Relevant: screen compound libraries for Parkinson's disease-relevant molecules

More Physiologically Relevant
The LanthaScreen™ Tb-Anti-LRRK2 [pSer935] Antibody allows the investigation of phosphorylation at serine 935 on LRRK2 protein kinase. When paired with LRRK2 BacMam reagents, you have the freedom to choose the cellular background for your assay, including primary cells. This enables screening for potential inhibitors of LRRK2 proteins in their natural complexes in a physiologically relevant cell type.

More Convenient
Assays are run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as western blotting and ELISA, making this assay ideal for HTS applications. The LanthaScreen™ technology provides all of the advantages of TR-FRET ratiometric detection, including reduced data noise, less interference from fluorescent compounds, and high sensitivity, which results in the use of fewer cells than traditional Western or ELISA methods.

Disease Relevant
The LanthaScreen™ Tb-Anti-LRRK2 [pSer935] Antibody can be used by Parkinson's and other disease researchers to perform HTS, cell-based assays to generate inhibition curves (IC50) for their compound libraries. This will aid drug discovery efforts by reducing the time to results from 4 hrs to typically 1.5 hrs compared to alternative ELISAs.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Related Links:
LRRK2 tools for advancing Parkinson's disease research
Kinase protein portfolio
LanthaScreen™ Eu Kinase Binding Assay
LanthaScreen™ Activity Assay
Learn More About BacMam Technology
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Cell-Based Assay
Assay Entry
Cell-based terbium modification activity (transient)
Cell State
Cells Not Included
Detection Method
Fluorescent
For Use With (Application)
TR-FRET
For Use With (Equipment)
Microplate Reader
Product Line
LanthaScreen™
Readout
TR-FRET (Activity)
Reporter Gene
GFP (EmGFP)
Target Entry
LRRK2 G2019S
Type
Cellular Assay Kit
System Type
LanthaScreen™
Druggable Target
Kinases

Contents & Storage

The kit includes:
• 5 tubes BacMam LRRK2-GFP G2019S 1 mL, Store at 4 °C
• 1 tube LanthaScreen™ 6X Lysis Buffer, Store at 4 °C
• 1 Tube LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody, Store at -20 °C
• 1 Instrument Control Tb TR-FRET Kit, Store at 4 °C
• 1 LanthaScreen™ LRRK2 [pSer935] G2019S Cellular Asasy Kit Protocol, Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text