Thermo Scientific Chemicals

Acetonyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A14691.14
25 g, Each
Thermo Scientific Chemicals

Acetonyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A14691.14
25 g, Each
Quantity
Catalog number: A14691.14
also known as A14691-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1235-21-8
IUPAC Name
(2-oxopropyl)triphenylphosphanium chloride
Molecular Formula
C21H20ClOP
InChI Key
XAMZZEBAJZJERT-UHFFFAOYSA-M
SMILES
[Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
242.0-248.0?C

Description

Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

Solubility
Soluble in hot water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text