Thermo Scientific Chemicals

Tricine, 98+%, Thermo Scientific Chemicals

Catalog number: A14695.22
100 g, Each
Thermo Scientific Chemicals

Tricine, 98+%, Thermo Scientific Chemicals

Catalog number: A14695.22
100 g, Each
Quantity
Catalog number: A14695.22
also known as A14695-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5704-04-1
IUPAC Name
2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular Formula
C6H13NO5
InChI Key
SEQKRHFRPICQDD-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)NCC(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Good's buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Good′s buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

Solubility
Soluble in water.

Notes
pH range: 7.8-8.8Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text