Thermo Scientific Chemicals

Cyclopentane, 95%, Thermo Scientific Chemicals

Catalog number: A14697.AE
100 mL, Each
Thermo Scientific Chemicals

Cyclopentane, 95%, Thermo Scientific Chemicals

Catalog number: A14697.AE
100 mL, Each
Quantity
Catalog number: A14697.AE
also known as A14697-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
287-92-3
IUPAC Name
cyclopentane
Molecular Formula
C5H10
InChI Key
RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES
C1CCCC1
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Refractive Index
1.4030-1.4090 @ 20?C
Form
Liquid

Description

Cyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.

Solubility
Miscible with ethanol, ether and acetone. Slightly miscible with water.

Notes
Avoid heat, direct sunlight, flames and sparks. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text