2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Robert D.Miller; Victor Y.Lee; Christopher R.Moylan. Substituted azole derivatives as nonlinear optical chromophores. Chem. Mater. 1994, 6, (7),1023-1032