Thermo Scientific Chemicals

4-n-Decyloxybenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A14703.14
25 g, Each
Thermo Scientific Chemicals

4-n-Decyloxybenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A14703.14
25 g, Each
Quantity
Catalog number: A14703.14
also known as A14703-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5519-23-3
IUPAC Name
4-(decyloxy)benzoic acid
Molecular Formula
C17H26O3
InChI Key
NZNICZRIRMGOFG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
Assay (Aqueous acid-base Titration)
≥95.0% to ≤105.0%
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥95.0%
Form
Powder

Description

4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,

Solubility
Soluble in water.

Notes
m.p. 97°(S), 122°(N), 142°(I)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text