Tetraphenylmethane, 96%, Thermo Scientific Chemicals
Tetraphenylmethane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraphenylmethane, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A14706.14
also known as A14706-14
Price (USD)/ Each
1,186.65
Online exclusive
1318.00 
Save 131.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,186.65
Online exclusive
1318.00 
Save 131.35 (10%)
Add to cart
Tetraphenylmethane, 96%, Thermo Scientific Chemicals
Catalog numberA14706.14
Price (USD)/ Each
1,186.65
Online exclusive
1318.00 
Save 131.35 (10%)
-
Add to cart
Chemical Identifiers
CAS630-76-2
IUPAC Name(triphenylmethyl)benzene
Molecular FormulaC25H20
InChI KeyPEQHIRFAKIASBK-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormPowder
Assay (HPLC)≥95.0%
Purity (DSC)≥95.0%
Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Brutschy, M.; Stangenberg, R.; Beer, C.; Lubczyk, D.; Baumgarten, M.; Müllen, K.; Waldvogel, S. R. The Generation Effect: Cavity Accessibility in Dense-Shell Polyphenylene Dendrimers. ChemPlusChem 2015, 80 (1), 54-56.
  2. Shen, K.; Zhang, Z.; Zhang, H.; Pang, J.; Jiang, Z. Poly(arylene ether ketone) carrying hyperquaternized pendants: Preparation, stability and conductivity. J. Power Sources 2015, 287, 432-440.