Appearance (Color)Colorless to white to pale yellow
FormFused solid or clear liquid as melt
Assay (GC)≥97.5%
Refractive Index1.5090-1.5130 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.