Thermo Scientific Chemicals

6-Aminohexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14719.36
500 g, Each
Thermo Scientific Chemicals

6-Aminohexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14719.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14719.36
also known as A14719-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-32-2
IUPAC Name
6-aminohexanoic acid
Molecular Formula
C6H13NO2
InChI Key
SLXKOJJOQWFEFD-UHFFFAOYSA-N
SMILES
NCCCCCC(O)=O
Appearance (Color)
White
Form
Crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5% to ≤101.5%
Identification (FTIR)
Conforms
Melting Point
ca. 200-210?C dec.

Description

6-Aminohexanoic acid is used as an intermediate in the polymerization of Nylon-6. It is involved in the extraction of aldehydes from the reaction mixture as well as to improve the solubilization of membrane proteins in electrophoresis. It acts as an inhibitor of fibrinolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 6-aminohexanoic acid, an epsilon-amino acid, shows an antifibrinolytic and hematologic activity
  • This compound competitively inhibits the activation of plasminogen; it consequently reduces the conversion of plasminogen to plasmin (fibrinolysin), an enzyme responsible for the degradation of fibrin clots, fibrinogen, and other plasma proteins, including the procoagulant factors V and VIII

Application

  • It has been used as an intermediate in the polymerization of Nylon-6
  • It is used for biomolecular applications to extract aldehydes from the reaction mixture and to increase membrane protein solubilization in electrophoresis experiments
  • It has been described to inhibit plasminogen binding to activated platelets
  • This compound can bind and inactivate carboxypeptidase B
  • This compound can also inhibit the activation of the first component of the complement system
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text