Thermo Scientific Chemicals

1-Bromooctane, 98+%, Thermo Scientific Chemicals

Catalog number: A14722.0B
1000 g, Each
Thermo Scientific Chemicals

1-Bromooctane, 98+%, Thermo Scientific Chemicals

Catalog number: A14722.0B
1000 g, Each
Quantity
Catalog number: A14722.0B
also known as A14722-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-83-1
IUPAC Name
1-bromooctane
Molecular Formula
C8H17Br
InChI Key
VMKOFRJSULQZRM-UHFFFAOYSA-N
SMILES
CCCCCCCCBr
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4505-1.4545 @ 20?C

Description

1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text