3-Bromobenzotrifluoride, 98+%, Thermo Scientific Chemicals
3-Bromobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A14724.36
also known as A14724-36
Price (USD)/ Each
414.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
414.00
Add to cart
3-Bromobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Catalog numberA14724.36
Price (USD)/ Each
414.00
-
Add to cart
Chemical Identifiers
CAS401-78-5
IUPAC Name1-bromo-3-(trifluoromethyl)benzene
Molecular FormulaC7H4BrF3
InChI KeyNNMBNYHMJRJUBC-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.4705-1.4745 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only