Thermo Scientific Chemicals

n-Decane, 99%, Thermo Scientific Chemicals

Catalog number: A14732.AP
500 mL, Each
Thermo Scientific Chemicals

n-Decane, 99%, Thermo Scientific Chemicals

Catalog number: A14732.AP
500 mL, Each
Quantity
Catalog number: A14732.AP
also known as A14732-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-18-5
IUPAC Name
decane
Molecular Formula
C10H22
InChI Key
DIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILES
CCCCCCCCCC
Refractive Index
1.4100-1.4130 @ 20?C (UK Sourced Material)
Assay (GC)
≥98.5%
Form
Liquid
Identification (FTIR)
Conforms (UK Sourced Material)
Appearance (Color)
Clear colorless

Description

n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

Solubility
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text