Thermo Scientific Chemicals

trans-Chalcone, 97%, Thermo Scientific Chemicals

Catalog number: A14734.22
100 g, Each
Thermo Scientific Chemicals

trans-Chalcone, 97%, Thermo Scientific Chemicals

Catalog number: A14734.22
100 g, Each
Quantity
Catalog number: A14734.22
also known as A14734-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-47-1
IUPAC Name
(2E)-1,3-diphenylprop-2-en-1-one
Molecular Formula
C15H12O
InChI Key
DQFBYFPFKXHELB-VAWYXSNFSA-N
SMILES
O=C(C=CC1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥96.0%
Appearance (Color)
Pale yellow to yellow
Melting Point (clear melt)
51.0-60.0?C
Form
Powder
Identification (FTIR)
Conforms

Description

Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text