Diethyl suberate, 99%, Thermo Scientific Chemicals
Diethyl suberate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl suberate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
500 g
Catalog number A14735.30
also known as A14735-30
Price (USD)/ Each
454.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
454.00
Add to cart
Diethyl suberate, 99%, Thermo Scientific Chemicals
Catalog numberA14735.30
Price (USD)/ Each
454.00
-
Add to cart
Chemical Identifiers
CAS2050-23-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4310-1.4340 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only