Thermo Scientific Chemicals

2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

Catalog number: A14741.0F
2500 mL, Each
Thermo Scientific Chemicals

2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

Catalog number: A14741.0F
2500 mL, Each
Quantity
Catalog number: A14741.0F
also known as A14741-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-60-8
IUPAC Name
2-methylcyclohexan-1-one
Molecular Formula
C7H12O
InChI Key
LFSAPCRASZRSKS-UHFFFAOYNA-N
SMILES
CC1CCCCC1=O
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4460-1.4500 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

Solubility
Insoluble in water. Soluble in methanol and diethyl ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text