Thermo Scientific Chemicals

Methyl propiolate, 99%, Thermo Scientific Chemicals

Catalog number: A14743.06
5 g, Each
Thermo Scientific Chemicals

Methyl propiolate, 99%, Thermo Scientific Chemicals

Catalog number: A14743.06
5 g, Each
Quantity
Catalog number: A14743.06
also known as A14743-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
922-67-8
IUPAC Name
methyl prop-2-ynoate
Molecular Formula
C4H4O2
InChI Key
IMAKHNTVDGLIRY-UHFFFAOYSA-N
SMILES
COC(=O)C#C
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4065-1.4095 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

Methyl propiolate was used in the synthesis of polysubstituted 3-arylaminoacrylate and tetrahydropyrimidin-2-one derivatives. It was also used as a thiol derivatizing agent for capillary electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl propiolate was used in the synthesis of polysubstituted 3-arylaminoacrylate and tetrahydropyrimidin-2-one derivatives. It was also used as a thiol derivatizing agent for capillary electrophoresis.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text