Thermo Scientific Chemicals

trans-3-(3-Pyridyl)acrylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14747.30
250 g, Each
Thermo Scientific Chemicals

trans-3-(3-Pyridyl)acrylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14747.30
250 g, Each
Quantity
Catalog number: A14747.30
also known as A14747-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19337-97-4
IUPAC Name
(2E)-3-(pyridin-3-yl)prop-2-enoic acid
Molecular Formula
C8H7NO2
InChI Key
VUVORVXMOLQFMO-ONEGZZNKSA-N
SMILES
OC(=O)C=CC1=CC=CN=C1
Form
Powder
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

Notes
Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text