Thermo Scientific Chemicals

4-Bromobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14755.22
100 g, Each
Thermo Scientific Chemicals

4-Bromobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14755.22
100 g, Each
Quantity
Catalog number: A14755.22
also known as A14755-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-90-4
IUPAC Name
(4-bromophenyl)(phenyl)methanone
Molecular Formula
C13H9BrO
InChI Key
KEOLYBMGRQYQTN-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Melting Point (clear melt)
78.0-84.0?C
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Powder

Description

4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

Solubility
Insoluble in water but slightly soluble in other solvents such as ethanol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text