Thermo Scientific Chemicals

2-Ethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14758.14
25 g, Each
Thermo Scientific Chemicals

2-Ethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14758.14
25 g, Each
Quantity
Catalog number: A14758.14
also known as A14758-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
134-11-2
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5385-1.5425 @ 20?C
Appearance (Color)
Clear colorless to white to yellow to pale brown
Form
Liquid or viscous liquid

Description

2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text