Thermo Scientific™

1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A14765.18
50 g, Each
Thermo Scientific™

1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A14765.18
50 g, Each
Quantity
Catalog number: A14765.18
also known as A14765-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-53-9
IUPAC Name
1,2-dibromobenzene
Molecular Formula
C6H4Br2
InChI Key
WQONPSCCEXUXTQ-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=C1Br
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.6090-1.6130 @ 20?C
Appearance (Color)
Clear colorless to yellow to pale brown

Description

1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text