Thermo Scientific Chemicals

Methyl 3-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A14766.06
5 g, Each
Thermo Scientific Chemicals

Methyl 3-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A14766.06
5 g, Each
Quantity
Catalog number: A14766.06
also known as A14766-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4518-10-9
IUPAC Name
methyl 3-aminobenzoate
Molecular Formula
C8H9NO2
InChI Key
VZDNXXPBYLGWOS-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(N)=C1
Appearance (Color)
Orange to red to pale brown to brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%

Description

Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water (0.1-1%) .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text