Thermo Scientific Chemicals

(R)-(-)-Mandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14767.14
25 g, Each
Thermo Scientific Chemicals

(R)-(-)-Mandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14767.14
25 g, Each
Quantity
Catalog number: A14767.14
also known as A14767-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-71-2
IUPAC Name
(2R)-2-hydroxy-2-phenylacetic acid
Molecular Formula
C8H8O3
InChI Key
IWYDHOAUDWTVEP-SSDOTTSWSA-N
SMILES
O[C@@H](C(O)=O)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or flakes
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms

Description

(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

Solubility
It is partly soluble in water, freely soluble in isopropyl and ethyl alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under ordinary conditions. Incompatabile to strong bases, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text