Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals
Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14768.14
also known as A14768-14
Price (USD)/ Each
224.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
224.00
Add to cart
Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals
Catalog numberA14768.14
Price (USD)/ Each
224.00
-
Add to cart
Chemical Identifiers
CAS39931-77-6
IUPAC Nameethyl 2-(pyridin-3-yl)acetate
Molecular FormulaC9H11NO2
InChI KeyRPWXYCRIAGBAGY-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4985-1.5015 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only