Thermo Scientific Chemicals

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14770.18
50 g, Each
Thermo Scientific Chemicals

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14770.18
50 g, Each
Quantity
Catalog number: A14770.18
also known as A14770-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2312-23-4
IUPAC Name
hydrogen (3-chlorophenyl)hydrazine chloride
Molecular Formula
C6H8Cl2N2
InChI Key
CRRIAWUJYMLJOE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=CC(Cl)=C1
Appearance (Color)
Cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text