Thermo Scientific Chemicals

3-(Methylthio)propionaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A14774.18
50 g, Each
Thermo Scientific Chemicals

3-(Methylthio)propionaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A14774.18
50 g, Each
Quantity
Catalog number: A14774.18
also known as A14774-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3268-49-3
IUPAC Name
3-(methylsulfanyl)propanal
Molecular Formula
C4H8OS
InChI Key
CLUWOWRTHNNBBU-UHFFFAOYSA-N
SMILES
CSCCC=O
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Free acid (titration)
≤1.5%

Description

3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text