Thermo Scientific Chemicals

4-Fluorostyrene, 97+%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A14775.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorostyrene, 97+%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A14775.14
25 g, Each
Quantity
Catalog number: A14775.14
also known as A14775-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
405-99-2
IUPAC Name
1-ethenyl-4-fluorobenzene
Molecular Formula
C8H7F
InChI Key
JWVTWJNGILGLAT-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C)C=C1
Refractive Index
1.5125-1.5175 @ 20?C
Solution Test
Clear (10% in Methanol)
Appearance (Color)
Clear colorless
Assay (GC)
>97.0%
Solution Test
Clear (10% in Diethyl ether)

Description

4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from acids, bases, oxidizing agents, halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text